Merck
  • Conformations of methyl 2,5-di-O-acetyl-beta-D-glucofuranosidurono-6,3-lactone and 1,2,5-tri-O-acetyl-beta-D-glucofuranurono-6,3-lactone in the crystal structure and in solution.
Conformations of methyl 2,5-di-O-acetyl-beta-D-glucofuranosidurono-6,3-lactone and 1,2,5-tri-O-acetyl-beta-D-glucofuranurono-6,3-lactone in the crystal structure and in solution.
Carbohydrate research (2006-07-15)
Beata Liberek, Dorota Tuwalska, Iwona do Santos-Zounon, Antoni Konitz, Artur Sikorski, Zygfryd Smiatacz
ABSTRACT
The single-crystal X-ray diffraction and high-resolution 1H and 13C NMR spectral data for methyl 2,5-di-O-acetyl-beta-D-glucofuranosidurono-6,3-lactone and 1,2,5-tri-O-acetyl-beta-D-glucofuranurono-6,3-lactone are reported. The lactones were synthesized as byproducts of reactions carried out to obtain methyl 1,2,3,4-tetra-O-acetyl-D-glucopyranuronate. The conformations of these lactones in the crystal structure and in solution are discussed. A 1T2-like conformation was found to be the preferred form for these lactones in both the crystal lattice and in solution.
Sigma-Aldrich
1,2,3,4-Tetra-O-acetyl-β-D-glucopyranose, 98%