Merck
  • Biocatalytic production of enantiopure cyclohexane-trans-1,2-diol using extracellular lipases from Bacillus subtilis.
Biocatalytic production of enantiopure cyclohexane-trans-1,2-diol using extracellular lipases from Bacillus subtilis.
Applied microbiology and biotechnology (2006-04-06)
Jean Detry, Thorsten Rosenbaum, Stephan Lütz, Doris Hahn, Karl-Erich Jaeger, Michael Müller, Thorsten Eggert
ABSTRACT
Two extracellular lipases from Bacillus subtilis, B. subtilis lipase A and lipase B, have been expressed in the heterologous host Escherichia coli, biochemically characterized and used for the kinetic resolution of (rac)-trans-1,2-diacetoxycyclohexane. Both enzymes were selectively acting on the (R,R)-enantiomer of the racemic substrate, highly specifically hydrolyzing only one of the two ester groups present, thus allowing the preparation of enantiopure (R,R)- and (S,S)-cyclohexane-trans-1,2-diol. The reaction conditions for the use of purified enzyme and crude cell lyophilizate were optimized and reactions in batch and repetitive batch modes were carried out on a preparative scale to yield enantiopure product (>99% enantiomeric excess).
Sigma-Aldrich
cis-1,2-Cyclohexanediol, 99%
Sigma-Aldrich
trans-1,2-Cyclohexanediol, 98%