- [Extravesical treatment of the ureter and endoscopic correction of reflux in children].
[Extravesical treatment of the ureter and endoscopic correction of reflux in children].
Vestnik khirurgii imeni I. I. Grekova (1991-03-01)
P P Salov, N S Zakharova
PMID1654647
ABSTRACT
The authors have suggested the presence of reflux regulating the relationship between the urinary bladder and vesicoureteral segment which was called the vesico-osteal reflex (VOR). An original method of diagnostics of VOR with the help of profilometry was developed. A method of endoscopic correction of reflux was proposed with the help of glue MK-2 and KL-3 in combination with metoclopramide++ which made the treatment considerably more effective.
Sigma-Aldrich
Poly(propylene glycol), tolylene 2,4-diisocyanate terminated, average Mn ~2,300 (narrow MW distribution), isocyanate ~3.6 wt. %