Merck
  • [Extraction of nitrobenzene and its metabolite 3-aminohydroxybenzene from water solutions].
[Extraction of nitrobenzene and its metabolite 3-aminohydroxybenzene from water solutions].
Sudebno-meditsinskaia ekspertiza (2006-03-03)
O I Malykhina, V K Shormanov
ABSTRACT
The article presents the results of nitrobenzene and 3-aminohydroxibensene extraction from aqueous solutions by means of five organic solvents. The following factors influence on the extraction degree is shown: the nature of the extracting agent, aqueous phase pH, saturation of some extracting agents with water. The number of extractions for isolation of the necessary quantities of the compounds studied is calculated.
Sigma-Aldrich
3-Aminophenol, 98%
Supelco
3-Aminophenol, PESTANAL®, analytical standard