Merck
  • Monitoring of antifouling booster biocides in water and sediment from the port of Osaka, Japan.
Monitoring of antifouling booster biocides in water and sediment from the port of Osaka, Japan.
Archives of environmental contamination and toxicology (2005-03-08)
Hiroya Harino, Yoshiaki Mori, Yoshitaka Yamaguchi, Kiyoshi Shibata, Tetsuya Senda
ABSTRACT
Concentrations of booster antifouling compounds in the port of Osaka, Japan were assessed. Concentrations of Sea-Nine 211 (4,5-dichloro-2-n-octyl-3-isothiazolone), thiabendazole (2-(4-thiazolyl)-benzimidazole), IPBC (3-iodo-2-propynyl butylcarbamate), Diuron (3,4-dichlorophenyl-N, N-dimethylurea), Irgarol 1051 (2-methylthio-4-t-butylamino-6-cyclopropylamino-s-triazine), and M1 (2-methylthio-4-t-butylamino-6-amino-s-triazine) in port water samples were in the range of <0.003-0.004 microg L(-1), <0.0008-0.020 microg L(-1), <0.0007-1.54 microg L(-1), <0.0008-0.267 microg L(-1), and <0.0019-0.167 microg L(-1), respectively. IPBC was not detected in the water samples, but the concentration of Diuron was higher than any previously reported. The concentrations of Sea-Nine 211, thiabendazole, Diuron, Irgarol 1051, and M1 in sediment samples were in the range of <0.04-2.4 microg kg(-1) dry, <0.08-1.2 microg kg(-1) dry, <0.64-1350 microg kg(-1) dry, <0.08-8.2 microg kg(-1) dry, and <0.18-2.9 microg kg(-1) dry, respectively. IPBC was again not detected. The levels of Sea-Nine 211, Diuron, and Irgarol 1051 in water and sediment samples were high in a poorly flushed mooring area for small and medium-hull vessels. Levels of Diuron and Irgarol 1051 were highest in summer. The concentration of Sea-Nine 211 in water increased between August and October 2002. Except for M1, increases in the levels of booster biocides in sediment were observed during the study period. The sediment-water partition (Kd) was calculated by dividing the concentrations in sediment by the concentrations in water. The Kd values for Sea-Nine 211, thiabendazole, Diuron, Irgarol 1051, and M1 were 690, 180, 2700, 300, and 870. The Kd value for these alternative compounds was lower than for TBT.
Sigma-Aldrich
3-Iodo-2-propynyl N-butylcarbamate, 97%