Merck
  • MMA/MPEOMA/VSA copolymer as a novel blood-compatible material: ex vivo platelet adhesion study.
MMA/MPEOMA/VSA copolymer as a novel blood-compatible material: ex vivo platelet adhesion study.
Journal of materials science. Materials in medicine (2004-08-28)
Jin Ho Lee, Se Heang Oh, Won Gon Kim
ABSTRACT
MMA/MPEOMA/VSA copolymers with both pendant polyethylene oxide (PEO) side chains and negatively chargeable side groups were synthesized by random copolymerization of methyl methacrylate (MMA), methoxy PEO monomethacrylate (MPEOMA; PEO mol. wt, 1000), and vinyl sulfonic acid sodium salt (VSA) monomers with different monomer composition to evaluate their blood compatibility. MMA/MPEOMA copolymer (with PEO side chains) and MMA/VSA copolymer (with negatively chargeable side groups) were also synthesized for the comparison purpose. The synthesized copolymers were coated onto polyurethane (PU) tubes (inner diameter, 4.6 mm) by a spin coating. The platelet adhesion of the MMA/MPEOMA/VSA copolymer-coated tube surfaces was compared with that of tube surface coated with MMA/MPEOMA or MMA/VSA copolymer with similar MPEOMA or VSA composition, using an ex vivo canine arterio-artery shunt method. The platelet adhesion was evaluated by radioactivity counting of technetium (99mTc)-labeled platelets adhered on the surfaces after 30 and 120 min of blood circulation. The MMA/MPEOMA/VSA copolymer (monomer molar ratio 9/0.5/0.5 or 8/1/1) was better in preventing platelet adhesion on the surface than the MMA/MPEOMA or MMA/VSA copolymer with similar MPEOMA or VSA composition, probably owing to the combined effects of highly mobile, hydrophilic PEO side chains and negatively charged VSA side groups.
Sigma-Aldrich
2-Amino-1-phenylethanol, 98%