Merck
  • Biomolecule-assisted synthesis of highly ordered snowflakelike structures of bismuth sulfide nanorods.
Biomolecule-assisted synthesis of highly ordered snowflakelike structures of bismuth sulfide nanorods.
Journal of the American Chemical Society (2004-01-08)
Qingyi Lu, Feng Gao, Sridhar Komarneni
ABSTRACT
A biomolecule-assisted simple technique has been developed for the spontaneous ordering of the Bi2S3 nanorods into snowflakelike superstructures in high yield under microwave-hydrothermal conditions. In this method, glutathione (GSH) is used as both an assembling agent and a sulfur source. By controlling the molar ratio between bismuth nitrate and glutathione as well as the synthetic temperature, several kinds of Bi2S3 one-dimensional nanomaterials such as snowflakelike structures, nanowires constructed of particles, short nanorods, and fine and long nanowires have been controllably synthesized.
Sigma-Aldrich
Bismuth(III) sulfide, 99%