Merck
  • Effect of ligand structure on the zinc-catalyzed Henry reaction. Asymmetric syntheses of (-)-denopamine and (-)-arbutamine.
Effect of ligand structure on the zinc-catalyzed Henry reaction. Asymmetric syntheses of (-)-denopamine and (-)-arbutamine.
Organic letters (2002-08-03)
Barry M Trost, Vince S C Yeh, Hisanako Ito, Nadine Bremeyer
ABSTRACT
[reaction: see text] Syntheses of variously modified ligands for the dinuclear zinc catalysts for the asymmetric aldol and nitroaldol (Henry) reactions are reported. Catalytic enantioselective nitroaldol reactions promoted by these modified ligands led to efficient syntheses of the beta-receptor agonists (-)-denopamine and (-)-arbutamine.
Sigma-Aldrich
R(−)-Denopamine, ≥98% (HPLC), powder