Merck
  • Factors that determine the plasma-membrane potential in bloodstream forms of Trypanosoma brucei.
Factors that determine the plasma-membrane potential in bloodstream forms of Trypanosoma brucei.
European journal of biochemistry (2000-07-21)
D P Nolan, H P Voorheis
ABSTRACT
The plasma-membrane potential (Delta(psi)p) in bloodstream forms of Trypanosoma brucei was studied using several different radiolabelled probes: 86Rb+ and [14C]SCN- were used to report Delta(psi)p directly because they distribute in easily measured quantities across the plasma membrane only, and [3H]methyltriphenylphosphonium (MePh3P+) was used to report Delta(psi)p only when Delta(psi)m had been abolished with FCCP because it reports the algebraic sum of the two potentials when used alone. The unperturbed Delta(psi)p had a value of -82 mV and was found to be essentially identical with, and determined almost completely by, the potassium diffusion potential, as evidenced by: (a) the lack of effect of valinomycin on the value obtained under appropriate conditions when any of these probes were used; (b) the close agreement of this measured value with that predicted from the measured distribution of K+ across the plasma membrane (-76 mV); (c) the large effect of changes in the extracellular K+ concentration by substitution with Na+ on Delta(psi)p together with the complete lack of effect of substitution of extracellular Na+ by the choline cation or substitution of extracellular Cl- by the gluconate anion on Delta(psi)p. The contribution to Delta(psi)p by electrogenic pumping of Na+/K+-ATPase was found to be small (of the order of 6 mV). H+ was not found to be pumped across the plasma membrane or to contribute to Delta(psi)p.
Sigma-Aldrich
Methyltriphenylphosphonium bromide, 98%