Merck
  • Covalent immobilization of unilamellar liposomes in gel beads for chromatography.
Covalent immobilization of unilamellar liposomes in gel beads for chromatography.
Analytical biochemistry (1999-03-17)
Q Yang, X Y Liu, M Yoshimoto, R Kuboi, J Miyake
ABSTRACT
For immobilized (proteo)liposome chromatography, unilamellar liposomes were covalently bound within gel beads that had been activated by CNBr, N-hydroxysuccinimide, tresyl, or chloroformate. Liposomes composed of phosphatidylcholine (PC) and 2 mol% of amino-containing lipid (phosphatidylethanolamine-caproylamine) were immobilized in the activated gels at 5-35 micromol lipid/ml gel and yields of 11-70%. The highest immobilized amount was found in chloroformate-activated TSK G6000PW gel, which contains large pore size (>100 nm). Liposomes composed of PC alone could also be attached to the chloroformate-activated gels at 33-42 micromol/ml gel and yields of 58-65%, probably by crosslinking of the phosphate moiety of phospholipid with the active group of the adsorbent. Liposomes prepared by various phospholipids with or without amino-containing lipids can generally be immobilized in the chloroformate-activated gels. The covalently bound liposomes were characterized by their high stability, unilamellarity, permeability of the membranes, and drug-membrane partition properties. A stable membrane phase was constructed for chromatographic experiments to be performed under extreme elution conditions.
Sigma-Aldrich
2,2,2-Trifluoroethanesulfonyl chloride, 99%