- Identification of the TRH-like peptides pGlu-Glu-Pro amide and pGlu-Phe-Pro amide in rat thyroid: regulation by thyroid status.
Identification of the TRH-like peptides pGlu-Glu-Pro amide and pGlu-Phe-Pro amide in rat thyroid: regulation by thyroid status.
Rat thyroid contains thyrotropin-releasing hormone (TRH) and TRH-like peptides which react with TRH antisera. We have identified the TRH-like peptides in the thyroid and examined whether their levels are influenced by thyroid status. The peptides were extracted from the thyroid glands of five hyperthyroid rats and purified by ion-exchange chromatography on SP-Sephadex C25 and reversed-phase high performance liquid chromatography. The principal TRH-immunoreactive component exhibited the same retention on HPLC as synthetic pGlu-Glu-Pro amide and a secondary component corresponded to synthetic pGlu-Phe-Pro amide. In agreement with these assignments the main peptide was shown to be acidic when chromatographed on DEAE-Sephadex A25 and the second peptide neutral. The levels of TRH and TRH-like peptides in the thyroid were investigated in hyper-, hypo- and euthyroid rats. Hyperthyroidism was induced by chronic subcutaneous administration of triiodothyronine (T3) and hypothyroidism was produced by addition of propylthiouracil (PTU) to the drinking water. The amounts of the peptides were determined by radioimmunoassay with a TRH-antiserum, carried out after extraction from the tissues and purification by ion exchange chromatography. The mean concentration of TRH-like peptides in the thyroids of the hyperthyroid rats was 95.5+/-25.5 pmol/g, the mean concentration in the hypothyroid rats was 11.7+/-3.4 pmol/g, and in the euthyroid rats 17.6+/-3.2 pmol/g. The concentrations of TRH were less influenced by thyroid status: the values in hyper-, hypo- and euthyroid rats were 47.5+/-9.4, 42.1+/-6.3, and 17.2+/-1.6 pmol/g respectively. The results show that the levels of the TRH-like peptides in rat thyroid are highly sensitive to thyroid status, suggesting a possible involvement in thyroid regulation.