Merck
Purification of Human Cytoplasmic Actins From Saccharomyces cerevisiae.
Bio-protocol (2023-12-14)
Brian K Haarer, David C Amberg, Jessica L Henty-Ridilla
ABSTRACT
Eukaryotic cells rely on actin to support cellular structure, motility, transport, and a wide variety of other cytoplasmic functions and nuclear activities. Humans and other mammals express six closely related isoforms of actin, four of which are found primarily in skeletal, cardiac, and smooth muscle tissues. The final two isoforms, β and γ, are found in non-muscle cells. Due to the ease of purification, many biochemical studies surveying the functions of actin and its regulators have been carried out with protein purified from skeletal muscle. However, it has become increasingly clear that some activities are isoform specific, necessitating more accessible sources of non-muscle actin isoforms. Recent innovations permit the purification of non-muscle actins from human cell culture and heterologous systems, such as insect cell culture and the yeast Pichia pastoris. However, these systems generate mixtures of actin types or require additional steps to remove purification-related tags. We have developed strains of Saccharomyces cerevisiae (budding yeast) that express single untagged isoforms of either human non-muscle actin (β or γ) as their sole actin, allowing the purification of individual homogeneous actin isoforms by conventional purification techniques. Key features • Easy growth of yeast as a source of human cytoplasmic actin isoforms. Uses well-established actin purification methods. • The tag-free system requires no post-purification processing.
Sigma-Aldrich
Phenylmethanesulfonyl fluoride, ≥98.5% (GC)
Millipore
Protease Inhibitor Cocktail Set IV, The Protease Inhibitor Cocktail Set IV controls the activity of Protease. This small molecule/inhibitor is primarily used for Protease Inhibitors applications.