Merck
  • Improving the accuracy of chloride measurements through participation in regular external quality assessment programme.
Improving the accuracy of chloride measurements through participation in regular external quality assessment programme.
Journal of trace elements in medicine and biology : organ of the Society for Minerals and Trace Elements (GMS) (2021-08-15)
Wesley Zongrong Yu, Qinde Liu, Ho Wah Leung, Benny M K Tong, Gina Chew, Tong Kooi Lee, Richard Y C Shin, Tang Lin Teo, Sunil Kumar Sethi
ABSTRACT
A chloride test is an integral part of a basic metabolic panel that is essential for the assessment of a patient's acid-base and electrolyte status. While many methods are available commercially for the routine measurement of chloride, there is a need to address the accuracy and variability among the measurement results, especially with the prevalence of patients seeking treatment across different healthcare providers for alternative opinions. A method based on sector field inductively coupled plasma isotope dilution mass spectrometry (SF-ICP-IDMS) was developed for the measurement of chloride in human serum. The SF-ICP-IDMS method was then used to assign the target values in the Health Sciences Authority (HSA) External Quality Assessment (EQA) Programme to evaluate the results of chloride test from participating clinical laboratories. The accuracy of the measurements was evaluated by comparing the results with the certified values of Electrolytes in Frozen Human Serum Certified Reference Materials (SRM 956c and SRM 956d) from the National Institute of Standards and Technology (NIST) at different chloride concentration levels. Over a five-year period from 2014-2018, the number of clinical laboratories which participated in the EQA Programme increased from 23 to 33. Comparison of robust means from the laboratories' results with our assigned target values revealed a reduction in relative deviation over time. The relationship between the deviation of each brand of clinical analysers and the chloride levels was established, where a larger deviation was uncovered at low chloride concentration. The SF-ICP-IDMS method was further demonstrated to be comparable with methods used by other metrology institutes in an international comparison organised by HSA under the auspice of the Consultative Committee for Amount of Substance - Metrology in Chemistry and Biology (CCQM). The use of metrologically traceable assigned target values enabled the study of method biasness from a small pool of dataset in each of the four brands of clinical analysers in HSA EQA Programme. This work underscores the need to improve the accuracy of chloride measurements by regular participation in an accuracy-based EQA Programme.