Merck
  • Achieving Nearly 30% External Quantum Efficiency for Orange-Red Organic Light Emitting Diodes by Employing Thermally Activated Delayed Fluorescence Emitters Composed of 1,8-Naphthalimide-Acridine Hybrids.
Achieving Nearly 30% External Quantum Efficiency for Orange-Red Organic Light Emitting Diodes by Employing Thermally Activated Delayed Fluorescence Emitters Composed of 1,8-Naphthalimide-Acridine Hybrids.
Advanced materials (Deerfield Beach, Fla.) (2017-12-09)
Weixuan Zeng, Hsin-Yu Lai, Wei-Kai Lee, Min Jiao, Yi-Jiun Shiu, Cheng Zhong, Shaolong Gong, Tao Zhou, Guohua Xie, Monima Sarma, Ken-Tsung Wong, Chung-Chih Wu, Chuluo Yang
ABSTRACT
The combination of rigid acridine donor and 1,8-naphthalimide acceptor has afforded two orange-red emitters of NAI-DMAC and NAI-DPAC with high rigidity in molecular structure and strongly pretwisted charge transfer state. Endowed with high photoluminescence quantum yields (ΦPL ), distinct thermally activated delayed fluorescence (TADF) characteristics, and preferentially horizontal emitting dipole orientations, these emitters afford record-high orange-red TADF organic light-emitting diodes (OLEDs) with external quantum efficiencies of up to 21-29.2%, significantly surpassing all previously reported orange-to-red TADF OLEDs. Notably, the influence of microcavity effect is verified to support the record-high efficiency. This finding relaxes the usually stringent material requirements for effective TADF emitters by comprising smaller radiative transition rates and less than ideal ΦPL s.
Sigma-Aldrich
4,4′-Bis(carbazol-9-yl)biphenyl, ≥99% (HPLC)
Pricing and availability is not currently available.
Sigma-Aldrich
Di-[4-(N,N-di-p-tolyl-amino)-phenyl]cyclohexane, ≥97% (HPLC)
Pricing and availability is not currently available.