Merck
  • Attenuation of Smad2 activity shows resistance to TGF-β signalling in mammary adenocarcinoma (MCF-7) cells.
Attenuation of Smad2 activity shows resistance to TGF-β signalling in mammary adenocarcinoma (MCF-7) cells.
Cell biology international (2013-03-16)
Suman Sengupta, Subhadip Kundu, Arindam Bhattacharyya
ABSTRACT
Transforming growth factor-β (TGF-β) is a potent inhibitor of the growth of normal mammary epithelial cells, and has a pleiotropic, context-dependent, concentration-dependent action. We found attenuation of TGF-β signalling in mammary adenoma carcinoma cells. Phosphorylation at the linker site of Smad2 occurred in a cooperative way during the attenuation of TGF-β signalling, and was associated with upregulation of CDK2 and cyclin D1. CDK2 inhibitor restored the anti-proliferative effect of TGF-β by upregulating p21, with inhibition of linker phosphorylation of Smad2. CDK2-mediated linker phosphorylation of Smad2 may be a plausible mechanism for the attenuation of TGF-β signalling in breast cancer.
Sigma-Aldrich
Goat Anti-Mouse IgG γ chain Antibody, FITC conjugate, Species Adsorbed, 1 mg/mL, Chemicon®