Merck
  • 2-piperidin-4-yl-benzimidazoles with broad spectrum antibacterial activities.
2-piperidin-4-yl-benzimidazoles with broad spectrum antibacterial activities.
Bioorganic & medicinal chemistry letters (2003-09-03)
Yun He, Baogen Wu, Jun Yang, Dale Robinson, Lisa Risen, Ray Ranken, Lawrence Blyn, Suzie Sheng, Eric E Swayze
ABSTRACT
A series of 2-piperidin-4-yl-benzimidazoles were synthesized and evaluated for antibacterial activities. Certain compounds inhibit bacterial growth with low micromolar minimal inhibitory concentration (MIC). These benzimidazoles are effective against both Gram-positive and Gram-negative bacteria of clinical importance, particularly enterococci, and represent a new class of potential antibacterial agents.
Sigma-Aldrich
Hydrogen chloride solution, 4.0 M in dioxane