Merck
  • Highly stable cesium lead iodide perovskite quantum dot light-emitting diodes.
Highly stable cesium lead iodide perovskite quantum dot light-emitting diodes.
Nanotechnology (2017-10-19)
Chen Zou, Chun-Ying Huang, Erin M Sanehira, Joseph M Luther, Lih Y Lin
ABSTRACT
Recently, all-inorganic perovskites such as CsPbBr3 and CsPbI3, have emerged as promising materials for light-emitting applications. While encouraging performance has been demonstrated, the stability issue of the red-emitting CsPbI3 is still a major concern due to its small tolerance factor. Here we report a highly stable CsPbI3 quantum dot (QD) light-emitting diode (LED) with red emission fabricated using an improved purification approach. The device achieved decent external quantum efficiency (EQE) of 0.21% at a bias of 6 V and outstanding operational stability, with a L 70 lifetime (EL intensity decreases to 70% of starting value) of 16 h and 1.5 h under a constant driving voltage of 5 V and 6 V (maximum EQE operation) respectively. Furthermore, the device can work under a higher voltage of 7 V (maximum luminance operation) and retain 50% of its initial EL intensity after 500 s. These findings demonstrate the promise of CsPbI3 QDs for stable red LEDs, and suggest the feasibility for electrically pumped perovskite lasers with further device optimizations.
Sigma-Aldrich
Lead(II) iodide, 99.999% trace metals basis