Merck
  • A cationic detergent, cetyltrimethylammonium bromide (CTAB), selectively dissociates the intermediate filament of the fibroblast.
A cationic detergent, cetyltrimethylammonium bromide (CTAB), selectively dissociates the intermediate filament of the fibroblast.
Biochemical and biophysical research communications (1991-03-29)
K Maki, J Sagara, A Kawai
ABSTRACT
We investigated conditions for selective solubilization of the intermediate filament (IF) of BHK-21 cells, and found that a cationic detergent, cetyltrimethylammonium bromide (CTAB), was effective for rapid dissociation of IF into the monomeric form. More selective dissociation was performed with a combination of CTAB and Tween 40. The CTAB-dissociated vimentins were unstable, but the breakdown of them was successfully blocked by leupeptin. Thus, with our extraction buffer, composed of 1% CTAB, 1% Tween 40, 10 mM Tris-HCl (pH 7.4) and 25 micrograms/ml leupeptin, almost all of the vimentin as well as the desmin were solubilized, while two thirds or more of actins were retained in the CTAB/Tween-insoluble fraction.
Sigma-Aldrich
Hexadecyltrimethylammonium bromide, ≥98%