Merck
  • Elucidating mechanisms of genetic cross-disease associations at the PROCR vascular disease locus.
Elucidating mechanisms of genetic cross-disease associations at the PROCR vascular disease locus.
Nature communications (2022-03-11)
David Stacey, Lingyan Chen, Paulina J Stanczyk, Joanna M M Howson, Amy M Mason, Stephen Burgess, Stephen MacDonald, Jonathan Langdown, Harriett McKinney, Kate Downes, Neda Farahi, James E Peters, Saonli Basu, James S Pankow, Weihong Tang, Nathan Pankratz, Maria Sabater-Lleal, Paul S de Vries, Nicholas L Smith, Amy D Gelinas, Daniel J Schneider, Nebojsa Janjic, Nilesh J Samani, Shu Ye, Charlotte Summers, Edwin R Chilvers, John Danesh, Dirk S Paul
ABSTRACT
Many individual genetic risk loci have been associated with multiple common human diseases. However, the molecular basis of this pleiotropy often remains unclear. We present an integrative approach to reveal the molecular mechanism underlying the PROCR locus, associated with lower coronary artery disease (CAD) risk but higher venous thromboembolism (VTE) risk. We identify PROCR-p.Ser219Gly as the likely causal variant at the locus and protein C as a causal factor. Using genetic analyses, human recall-by-genotype and in vitro experimentation, we demonstrate that PROCR-219Gly increases plasma levels of (activated) protein C through endothelial protein C receptor (EPCR) ectodomain shedding in endothelial cells, attenuating leukocyte-endothelial cell adhesion and vascular inflammation. We also associate PROCR-219Gly with an increased pro-thrombotic state via coagulation factor VII, a ligand of EPCR. Our study, which links PROCR-219Gly to CAD through anti-inflammatory mechanisms and to VTE through pro-thrombotic mechanisms, provides a framework to reveal the mechanisms underlying similar cross-phenotype associations.
Sigma-Aldrich
Protein C from human plasma, Activated, lyophilized powder, ≥90% (SDS-PAGE)