Merck
  • Applications of peptide hormone ligands for the treatment of dumping and short bowel syndrome.
Applications of peptide hormone ligands for the treatment of dumping and short bowel syndrome.
Current opinion in pharmacology (2018-10-03)
Lucas Wauters, Tim Vanuytsel
ABSTRACT
Dumping syndrome is a common and debilitating complication of upper gastrointestinal surgery. Accelerated gastric emptying and dysregulated secretion of gastrointestinal (GI) hormones are involved in its pathophysiology. Pasireotide, a novel somatostatin analogue, improved dumping in a phase-2 study. Preliminary data suggest that the glucagon-like peptide-1 (GLP-1) analogue liraglutide can also improve dumping. Short bowel syndrome is the most common cause of intestinal failure and involves not only a loss of mucosal absorptive area but also hypersecretion and accelerated transit. GLP-2 is the best studied hormone involved in intestinal adaptation. An increasing body of evidence demonstrates that the GLP-2 analogue teduglutide reduces parenteral support needs. New GLP-2 analogues and analogues of other GI hormones such as liraglutide are being investigated as promising treatments in short bowel syndrome.
Sigma-Aldrich
Teduglutide trifluoroacetate, ≥95% (HPLC)