Merck
  • Autoantibodies against appetite-regulating peptide hormones and neuropeptides: putative modulation by gut microflora.
Autoantibodies against appetite-regulating peptide hormones and neuropeptides: putative modulation by gut microflora.
Nutrition (Burbank, Los Angeles County, Calif.) (2008-02-12)
Sergueï O Fetissov, Maria Hamze Sinno, Moïse Coëffier, Christine Bole-Feysot, Philippe Ducrotté, Tomas Hökfelt, Pierre Déchelotte
ABSTRACT
Peptide hormones synthesized in gastrointestinal and adipose tissues in addition to neuropeptides regulate appetite and body weight. Previously, autoantibodies directed against melanocortin peptides were found in patients with eating disorders; however, it remains unknown whether autoantibodies directed against other appetite-regulating peptides are present in human sera and whether their levels are influenced by gut-related antigens. Healthy women were studied for the presence of immunoglobulin (Ig) G and IgA autoantibodies directed against 14 key appetite-regulating peptides. The concept of molecular mimicry was applied to search in silico whether bacteria, viruses, or fungi contain proteins with amino acid sequences identical to appetite-regulating peptides. In addition, autoantibodies serum levels were studied in germ-free and specific pathogen-free rats. We found these IgG and IgA autoantibodies directed against leptin, ghrelin, peptide YY, neuropeptide Y, and other appetite-regulating peptides are present in human sera at levels of 100-900 ng/mL. Numerous cases of sequence homology with these peptides were identified among commensal and pathogenic micro-organisms including Lactobacilli, bacteroides, Helicobacter pylori, Escherichia coli, and Candida species. Decreased levels of IgA autoantibodies directed against several appetite-regulating peptides and increased levels of antighrelin IgG were found in germ-free rats compared with specific pathogen-free rats. Healthy humans and rats display autoantibodies directed against appetite-regulating peptide hormones and neuropeptides, suggesting that these autoantibodies may have physiologic implications in hunger and satiety pathways. Gut-related antigens including the intestinal microflora may influence production of theses autoantibodies, suggesting a new link between the gut and appetite control.
Product Description
Millipore
Protein G–Agarose, lyophilized powder, Contains lactose stabilizers that must be removed prior to use.