Merck
  • A chemokine regulatory loop induces cholesterol synthesis in lung-colonizing triple-negative breast cancer cells to fuel metastatic growth.
A chemokine regulatory loop induces cholesterol synthesis in lung-colonizing triple-negative breast cancer cells to fuel metastatic growth.
Molecular therapy : the journal of the American Society of Gene Therapy (2021-07-19)
Bingchen Han, Felix Alonso-Valenteen, Zhe Wang, Nan Deng, Tian-Yu Lee, Bowen Gao, Ying Zhang, Yali Xu, Xinfeng Zhang, Sandrine Billet, Xuemo Fan, Stephen Shiao, Neil Bhowmick, Lali Medina-Kauwe, Armando Giuliano, Xiaojiang Cui
ABSTRACT
Triple-negative breast cancer (TNBC) has a high propensity for organ-specific metastasis. However, the underlying mechanisms are not well understood. Here we show that the primary TNBC tumor-derived C-X-C motif chemokines 1/2/8 (CXCL1/2/8) stimulate lung-resident fibroblasts to produce the C-C motif chemokines 2/7 (CCL2/7), which, in turn, activate cholesterol synthesis in lung-colonizing TNBC cells and induce angiogenesis at lung metastatic sites. Inhibiting cholesterol synthesis in lung-colonizing breast tumor cells by pulmonary administration of simvastatin-carrying HER3-targeting nanoparticles reduces angiogenesis and growth of lung metastases in a syngeneic TNBC mouse model. Our findings reveal a novel, chemokine-regulated mechanism for the cholesterol synthesis pathway and a critical role of metastatic site-specific cholesterol synthesis in the pulmonary tropism of TNBC metastasis. The study has implications for the unresolved epidemiological observation that use of cholesterol-lowering drugs has no effect on breast cancer incidence but can unexpectedly reduce breast cancer mortality, suggesting interventions of cholesterol synthesis in lung metastases as an effective treatment to improve survival in individuals with TNBC.
Sigma-Aldrich
FTI-277 trifluoroacetate salt, ≥95% (HPLC), film
Sigma-Aldrich
QCM 3 µm Endothelial Cell Migration Assay Fibronectin, Colorimetric