Merck
  • Secretory function of ovarian cells and myometrial contractions in cow are affected by chlorinated insecticides (chlordane, heptachlor, mirex) in vitro.
Secretory function of ovarian cells and myometrial contractions in cow are affected by chlorinated insecticides (chlordane, heptachlor, mirex) in vitro.
Toxicology and applied pharmacology (2016-11-27)
Michael Hubert Wrobel, Jaroslaw Mlynarczuk
ABSTRACT
The aim of the study was to investigate the effect of chlordane, heptachlor and mirex, on hormonal regulation of the force of myometrial contractions. Myometrial, endometrial, granulosa and luteal cells as well as strips of myometrium from non-pregnant cows were incubated with three insecticides at environmentally relevant doses (0.1, 1 or 10ng/ml). None of the insecticides affected the viability of studied cells. Chlordane stimulated, while heptachlor and mirex inhibited, secretion of testosterone and estradiol from granulosa cells as well as secretion of progesterone from luteal cells, respectively. Secretion of oxytocin (OT) from granulosa cells was increased after incubation with all studied insecticides. Only mirex stimulated OT secretion from luteal cells, while heptachlor inhibited this effect. None of them affected synthesis of OT in luteal cells and prostaglandins (PGF2 and PGE2) secretion from uterine cells, except PGE2 secretion from endometrial cells was decreased when the cells were incubated with 0.1ng/ml of chlordane. Basal and OT-stimulated myometrial contractions were increased by mirex and decreased by heptachlor. The data show that the insecticides altered secretory function of ovarian cells. Heptachlor and mirex affected also myometrial contractions in vitro, but uterine secretion of prostaglandins were not involved in the mechanism of that adverse effect of insecticides. The data indicate on potential of these insecticides to disturb fertilisation, blastocyst implantation or even the length of gestation.