Merck
  • Decreased arthritis severity in cathepsin L-deficient mice is attributed to an impaired T helper cell compartment.
Decreased arthritis severity in cathepsin L-deficient mice is attributed to an impaired T helper cell compartment.
Inflammation research : official journal of the European Histamine Research Society ... [et al.] (2012-06-08)
Uta Schurigt, Rene Eilenstein, Mieczyslaw Gajda, Carola Leipner, Lisa Sevenich, Thomas Reinheckel, Christoph Peters, Bernd Wiederanders, Rolf Bräuer
ABSTRACT
Cathepsin L (CL) is potentially involved in joint destruction and in antigen presentation in rheumatoid arthritis. In order to define the roles of this protease in arthritis development we analysed the antigen-induced arthritis (AIA) in CL-deficient (CL(-/-)) mice. Antigen-induced arthritis was induced in CL(-/-) and wild-type mice. Complete CL deficiency resulted in an impaired positive selection of conventional CD4(+) T helper (Th) cells and finally in a reduced number of Th cells. Thus, we addressed the effect of this phenotype by rescuing CD4(+) Th cell numbers by transgenic expression of the human CL-like protease cathepsin V (hCV) in thymic epithelium of CL(-/-) mice [Tg(K14-hCV);CL(-/-)]. The arthritis development was monitored by measuring joint swelling. Joint inflammation and destruction were assessed histopathologically. The severity of AIA was decreased in CL(-/-) mice characterized by reduced swelling, decreased inflammation and destruction, and diminished cellular and humoral immune responsiveness. AIA in Tg(K14-hCV);CL(-/-) mice was associated with a reconstitution of all parameters by normalization of the ratio of regulatory to conventional T cells. Cathepsin L has a significant impact on AIA severity by influencing the selection of Th cell populations in the thymus, but seems not play any significant role in the direct joint destruction.
Sigma-Aldrich
Cathepsin L Active human, recombinant, expressed in FreeStyle 293-F cells, ≥90% (SDS-PAGE)
Sigma-Aldrich
Cathepsin V Active human, recombinant, expressed in FreeStyle 293-F cells, ≥90% (SDS-PAGE)