Merck
  • Reelin Depletion Protects Against Atherosclerosis by Decreasing Vascular Adhesion of Leukocytes.
Reelin Depletion Protects Against Atherosclerosis by Decreasing Vascular Adhesion of Leukocytes.
Arteriosclerosis, thrombosis, and vascular biology (2021-02-26)
Laurent Calvier, Xunde Xian, Richard G Lee, Anastasia Sacharidou, Chieko Mineo, Philip W Shaul, Maria Z Kounnas, Shirling Tsai, Joachim Herz
ABSTRACT
[Figure: see text].
Sigma-Aldrich
Anti-Reelin Antibody, a.a. 164-189 mreelin, clone 142, clone 142, Chemicon®, from mouse
Sigma-Aldrich
Anti-GAPDH antibody, Mouse monoclonal, clone GAPDH-71.1, purified from hybridoma cell culture
Sigma-Aldrich
Anti-PI3 Kinase Antibody, p85, N-SH3, clone AB6, clone AB6, Upstate®, from mouse