Merck
  • Synergistic Photothermal-Chemotherapy Based on the Use of Biomimetic Magnetic Nanoparticles.
Synergistic Photothermal-Chemotherapy Based on the Use of Biomimetic Magnetic Nanoparticles.
Pharmaceutics (2021-05-01)
Ylenia Jabalera, Alberto Sola-Leyva, María P Carrasco-Jiménez, Guillermo R Iglesias, Concepcion Jimenez-Lopez
ABSTRACT
MamC-mediated biomimetic magnetic nanoparticles (BMNPs) have emerged as one of the most promising nanomaterials due to their magnetic features (superparamagnetic character and large magnetic moment per particle), their novel surface properties determined by MamC, their biocompatibility and their ability as magnetic hyperthermia agents. However, the current clinical application of magnetic hyperthermia is limited due to the fact that, in order to be able to reach an effective temperature at the target site, relatively high nanoparticle concentration, as well as high magnetic field strength and/or AC frequency are needed. In the present study, the potential of BMNPs to increase the temperature upon irradiation of a laser beam in the near infrared, at a wavelength at which tissues become partially transparent, is explored. Moreover, our results also demonstrate the synergy between photothermia and chemotherapy in terms of drug release and cytotoxicity, by using BMNPs functionalized with doxorubicin, and the effectiveness of this combination therapy against tumor cells in in vitro experiments. Therefore, the findings of the present study open the possibility of a novel, alternative approach to fight localized tumors.
Sigma-Aldrich
Resazurin sodium salt, powder, BioReagent