Merck
  • An Alu-like RNA promotes cell differentiation and reduces malignancy of human neuroblastoma cells.
An Alu-like RNA promotes cell differentiation and reduces malignancy of human neuroblastoma cells.
FASEB journal : official publication of the Federation of American Societies for Experimental Biology (2010-06-29)
Manuele Castelnuovo, Sara Massone, Roberta Tasso, Gloria Fiorino, Monica Gatti, Mauro Robello, Elena Gatta, Audrey Berger, Katharina Strub, Tullio Florio, Giorgio Dieci, Ranieri Cancedda, Aldo Pagano
ABSTRACT
Neuroblastoma (NB) is a pediatric cancer characterized by remarkable cell heterogeneity within the tumor nodules. Here, we demonstrate that the synthesis of a pol III-transcribed noncoding (nc) RNA (NDM29) strongly restricts NB development by promoting cell differentiation, a drop of malignancy processes, and a dramatic reduction of the tumor initiating cell (TIC) fraction in the NB cell population. Notably, the overexpression of NDM29 also confers to malignant NB cells an unpredicted susceptibility to the effects of antiblastic drugs used in NB therapy. Altogether, these results suggest the induction of NDM29 expression as possible treatment to increase cancer cells vulnerability to therapeutics and the measure of its synthesis in NB explants as prognostic factor of this cancer type.
Product Description
Supelco
Poly(ethyleneimine) solution, analytical standard, 50 % (w/v) in H2O