Merck
  • Fabrication of 3D Micro-Blades for the Cutting of Biological Structures in a Microfluidic Guillotine.
Fabrication of 3D Micro-Blades for the Cutting of Biological Structures in a Microfluidic Guillotine.
Micromachines (2021-09-29)
Saisneha Koppaka, Kevin S Zhang, Myra Kurosu Jalil, Lucas R Blauch, Sindy K Y Tang
ABSTRACT
Micro-blade design is an important factor in the cutting of single cells and other biological structures. This paper describes the fabrication process of three-dimensional (3D) micro-blades for the cutting of single cells in a microfluidic "guillotine" intended for fundamental wound repair and regeneration studies. Our microfluidic guillotine consists of a fixed 3D micro-blade centered in a microchannel to bisect cells flowing through. We show that the Nanoscribe two-photon polymerization direct laser writing system is capable of fabricating complex 3D micro-blade geometries. However, structures made of the Nanoscribe IP-S resin have low adhesion to silicon, and they tend to peel off from the substrate after at most two times of replica molding in poly(dimethylsiloxane) (PDMS). Our work demonstrates that the use of a secondary mold replicates Nanoscribe-printed features faithfully for at least 10 iterations. Finally, we show that complex micro-blade features can generate different degrees of cell wounding and cell survival rates compared with simple blades possessing a vertical cutting edge fabricated with conventional 2.5D photolithography. Our work lays the foundation for future applications in single cell analyses, wound repair and regeneration studies, as well as investigations of the physics of cutting and the interaction between the micro-blade and biological structures.
Sigma-Aldrich
3-(Trimethoxysilyl)propyl methacrylate, ≥97%