Merck
  • Glutamic acid gamma-monohydroxamate and hydroxylamine are alternate substrates for Escherichia coli asparagine synthetase B.
Glutamic acid gamma-monohydroxamate and hydroxylamine are alternate substrates for Escherichia coli asparagine synthetase B.
Biochemistry (1996-03-05)
S K Boehlein, S M Schuster, N G Richards
ABSTRACT
Escherichia coli asparagine synthetase B (AS-B) catalyzes the synthesis of asparagine from aspartic acid and glutamine in an ATP-dependent reaction. The ability of this enzyme to employ hydroxylamine and L-glutamic acid gamma-monohydroxamate (LGH) as alternative substrates in place of ammonia and L-glutamine, respectively, has been investigated. The enzyme is able to function as an amidohydrolase, liberating hydroxylamine from LGH with high catalytic efficiency, as measured by k(cat)/K(M). In addition, the kinetic parameters determined for hydroxylamine in AS-B synthetase activity are very similar to those of ammonia. Nitrogen transfer from LGH to yield aspartic acid beta-monohydroxamate is also catalyzed by AS-B. While such an observation has been made for a few members of the trpG amidotransferase family, our results appear to be the first demonstration that nitrogen transfer can occur from glutamine analogs in a purF amidotransferase. However, k(cat)/K(M) for the ATP-dependent transfer of hydroxylamine from LGH to aspartic acid is reduced 3-fold relative to that for glutamine-dependent asparagine synthesis. Further, the AS-B mutant in which asparagine is replaced by alanine (N74A) can also use hydroxylamine as an alternate substrate to ammonia and catalyze the hydrolysis of LGH. The catalytic efficiencies (k(cat)/K(M)) of nitrogen transfer from LGH and L-glutamine to beta-aspartyl-AMP are almost identical for the N74A AS-B mutant. These observations support the proposal that Asn-74 plays a role in catalyzing glutamine-dependent nitrogen transfer. We interpret our kinetic data as further evidence against ammonia-mediated nitrogen transfer from glutamine in the purF amidotransferase AS-B. These results are consistent with two alternate chemical mechanisms that have been proposed for this reaction [Boehlein, S. K., Richards, N. G. J., Walworth, E. S., & Schuster, S. M. (1994) J. Biol. Chem. 269, 26789-26795].
Sigma-Aldrich
L-Glutamic acid γ-monohydroxamate