Merck
  • Autoantibodies neutralizing type I IFNs are present in ~4% of uninfected individuals over 70 years old and account for ~20% of COVID-19 deaths.
Autoantibodies neutralizing type I IFNs are present in ~4% of uninfected individuals over 70 years old and account for ~20% of COVID-19 deaths.
Science immunology (2021-08-21)
Paul Bastard, Adrian Gervais, Tom Le Voyer, Jérémie Rosain, Quentin Philippot, Jérémy Manry, Eleftherios Michailidis, Hans-Heinrich Hoffmann, Shohei Eto, Marina Garcia-Prat, Lucy Bizien, Alba Parra-Martínez, Rui Yang, Liis Haljasmägi, Mélanie Migaud, Karita Särekannu, Julia Maslovskaja, Nicolas de Prost, Yacine Tandjaoui-Lambiotte, Charles-Edouard Luyt, Blanca Amador-Borrero, Alexandre Gaudet, Julien Poissy, Pascal Morel, Pascale Richard, Fabrice Cognasse, Jesus Troya, Sophie Trouillet-Assant, Alexandre Belot, Kahina Saker, Pierre Garçon, Jacques G Rivière, Jean-Christophe Lagier, Stéphanie Gentile, Lindsey B Rosen, Elana Shaw, Tomohiro Morio, Junko Tanaka, David Dalmau, Pierre-Louis Tharaux, Damien Sene, Alain Stepanian, Bruno Megarbane, Vasiliki Triantafyllia, Arnaud Fekkar, James R Heath, José Luis Franco, Juan-Manuel Anaya, Jordi Solé-Violán, Luisa Imberti, Andrea Biondi, Paolo Bonfanti, Riccardo Castagnoli, Ottavia M Delmonte, Yu Zhang, Andrew L Snow, Steven M Holland, Catherine Biggs, Marcela Moncada-Vélez, Andrés Augusto Arias, Lazaro Lorenzo, Soraya Boucherit, Boubacar Coulibaly, Dany Anglicheau, Anna M Planas, Filomeen Haerynck, Sotirija Duvlis, Robert L Nussbaum, Tayfun Ozcelik, Sevgi Keles, Ahmed A Bousfiha, Jalila El Bakkouri, Carolina Ramirez-Santana, Stéphane Paul, Qiang Pan-Hammarström, Lennart Hammarström, Annabelle Dupont, Alina Kurolap, Christine N Metz, Alessandro Aiuti, Giorgio Casari, Vito Lampasona, Fabio Ciceri, Lucila A Barreiros, Elena Dominguez-Garrido, Mateus Vidigal, Mayana Zatz, Diederik van de Beek, Sabina Sahanic, Ivan Tancevski, Yurii Stepanovskyy, Oksana Boyarchuk, Yoko Nukui, Miyuki Tsumura, Loreto Vidaur, Stuart G Tangye, Sonia Burrel, Darragh Duffy, Lluis Quintana-Murci, Adam Klocperk
ABSTRACT
Circulating autoantibodies (auto-Abs) neutralizing high concentrations (10 ng/mL, in plasma diluted 1 to 10) of IFN-α and/or -ω are found in about 10% of patients with critical COVID-19 pneumonia, but not in subjects with asymptomatic infections. We detect auto-Abs neutralizing 100-fold lower, more physiological, concentrations of IFN-α and/or -ω (100 pg/mL, in 1/10 dilutions of plasma) in 13.6% of 3,595 patients with critical COVID-19, including 21% of 374 patients > 80 years, and 6.5% of 522 patients with severe COVID-19. These antibodies are also detected in 18% of the 1,124 deceased patients (aged 20 days-99 years; mean: 70 years). Moreover, another 1.3% of patients with critical COVID-19 and 0.9% of the deceased patients have auto-Abs neutralizing high concentrations of IFN-β. We also show, in a sample of 34,159 uninfected subjects from the general population, that auto-Abs neutralizing high concentrations of IFN-α and/or -ω are present in 0.18% of individuals between 18 and 69 years, 1.1% between 70 and 79 years, and 3.4% >80 years. Moreover, the proportion of subjects carrying auto-Abs neutralizing lower concentrations is greater in a subsample of 10,778 uninfected individuals: 1% of individuals <70 years, 2.3% between 70 and 80 years, and 6.3% >80 years. By contrast, auto-Abs neutralizing IFN-β do not become more frequent with age. Auto-Abs neutralizing type I IFNs predate SARS-CoV-2 infection and sharply increase in prevalence after the age of 70 years. They account for about 20% of both critical COVID-19 cases in the over-80s, and total fatal COVID-19 cases.
Sigma-Aldrich
IFN-omega human, recombinant, expressed in E. coli, ≥98% (SDS-PAGE), ≥98% (HPLC), suitable for cell culture