Merck
  • Inhibition of beta-amyloid(40) fibrillogenesis and disassembly of beta-amyloid(40) fibrils by short beta-amyloid congeners containing N-methyl amino acids at alternate residues.
Inhibition of beta-amyloid(40) fibrillogenesis and disassembly of beta-amyloid(40) fibrils by short beta-amyloid congeners containing N-methyl amino acids at alternate residues.
Biochemistry (2001-07-11)
D J Gordon, K L Sciarretta, S C Meredith
ABSTRACT
A potential goal in the prevention or therapy of Alzheimer's disease is to decrease or eliminate neuritic plaques composed of fibrillar beta-amyloid (Abeta). In this paper we describe N-methyl amino acid containing congeners of the hydrophobic "core domain" of Abeta that inhibit the fibrillogenesis of full-length Abeta. These peptides also disassemble preformed fibrils of full-length Abeta. A key feature of the inhibitor peptides is that they contain N-methyl amino acids in alternating positions of the sequence. The most potent of these inhibitors, termed Abeta16-22m, has the sequence NH(2)-K(Me-L)V(Me-F)F(Me-A)E-CONH(2). In contrast, a peptide, NH(2)-KL(Me-V)(Me-F)(Me-F)(Me-A)-E-CONH(2), with N-methyl amino acids in consecutive order, is not a fibrillogenesis inhibitor. Another peptide containing alternating N-methyl amino acids but based on the sequence of a different fibril-forming protein, the human prion protein, is also not an inhibitor of Abeta40 fibrillogenesis. The nonmethylated version of the inhibitor peptide, NH(2)-KLVFFAE-CONH(2) (Abeta16-22), is a weak fibrillogenesis inhibitor. Perhaps contrary to expectations, the Abeta16-22m peptide is highly soluble in aqueous media, and concentrations in excess of 40 mg/mL can be obtained in buffers of physiological pH and ionic strength, compared to only 2 mg/mL for Abeta16-22. Analytical ultracentrifugation demonstrates that Abeta16-22m is monomeric in buffer solution. Whereas Abeta16-22 is susceptible to cleavage by chymotrypsin, the methylated inhibitor peptide Abeta16-22m is completely resistant to this protease. Circular dichroic spectroscopy of Abeta16-22m indicates that this peptide is a beta-strand, albeit with an unusual minimum at 226 nm. In summary, the inhibitor motif is that of alternating N-methyl and nonmethylated amino acids in a sequence critical for Abeta40 fibrillogenesis. These inhibitors appear to act by binding to growth sites of Abeta nuclei and/or fibrils and preventing the propagation of the network of hydrogen bonds that is essential for the formation of an extended beta-sheet fibril.
Sigma-Aldrich
Nα-Methyl-L-lysine monohydrochloride, ≥98.0% (TLC)
Sigma-Aldrich
Nα-Methyl-L-ornithine monohydrochloride, ≥98% (TLC)