Merck
  • Fabrication of 2D and 3D constructs from reconstituted decellularized tissue extracellular matrices.
Fabrication of 2D and 3D constructs from reconstituted decellularized tissue extracellular matrices.
Journal of biomedical nanotechnology (2015-05-24)
Yuji S Takeda, Qiaobing Xu
ABSTRACT
We demonstrated a novel process to reconstitute a decellularized extracellular matrix (Recon-ECM) of heart and liver tissue using a combination of mechanical homogenization and enzymatic digestion. Such Recon-ECM was used as a biomaterial to produce flat or micro-patterned 2D films after crosslinking using replica molding. The mechanical properties of the resulting films were tuned by changing the type of crosslinking reagents. We also demonstrated the fabrication of mechanically robust 3D scaffolds by freeze-drying of the Recon-ECM solution. The porosity of the 3D scaffold was controlled by changing the concentration of the Recon-ECM. HepG2 cells were used to investigate the potential substrate of these engineered 2D patterned and 3D porous structures. The cell attachment, proliferation, and urea synthesis were evaluated, and the results indicate that the scaffold generated from Recon-ECM provides a biologically friendly environment for cells to grow. This method provides a new way to use decellularized ECM as a source of biomaterial to produce novel scaffolds with better controlled micro- and nano-scale structures, tunable physicochemical properties with desired biological functions.
Product Description
Millipore
ReBlot Plus Kit