Merck
  • Effect of source of trace minerals in either forage- or by-product-based diets fed to dairy cows: 2. Apparent absorption and retention of minerals.
Effect of source of trace minerals in either forage- or by-product-based diets fed to dairy cows: 2. Apparent absorption and retention of minerals.
Journal of dairy science (2017-05-01)
M J Faulkner, N R St-Pierre, W P Weiss
ABSTRACT
Eighteen multiparous cows were used in a split-plot replicated Latin square with two 28-d periods to evaluate the effects of source of supplemental Cu, Zn, and Mn (sulfates or hydroxy) on apparent absorption of minerals when fed in either a forage- or by-product-based diet. The by-product diets were formulated to have greater concentrations of NDF and lesser concentrations of starch, and specific ingredients were chosen because they were good sources of soluble fiber and β-glucans, which bind trace minerals in nonruminants. We hypothesized that hydroxy trace minerals would interact less with digesta and have greater apparent absorption compared with sulfate minerals, and the difference in apparent absorption would be greater for the by-product diet compared with the forage-based diet. During the 56-d experiment, cows remained on the same fiber treatment but source of supplemental trace mineral was different for each 28-d period; thus, all cows were exposed to both mineral treatments. During each period cows were fed no supplemental Cu, Zn, or Mn for 16 d, followed by 12 d of feeding supplemental minerals from either sulfate or hydroxy sources. Supplemental minerals for each of the mineral sources fed provided approximately 10, 35, and 32 mg/kg of supplemental Cu, Zn, and Mn, respectively, for both fiber treatments. Total Cu, Zn, and Mn dietary concentrations, respectively, were approximately 19, 65, and 70 mg/kg for the forage diets and 21, 85, and 79 for the by-product diets. Treatment had no effect on dry matter intake (24.2 kg/d) or milk production (34.9 kg/d). Cows consuming the by-product diets had greater Zn (1,863 vs. 1,453 mg/d) and Mn (1,790 vs. 1,588 mg/d) intake compared with cows fed forage diets, but apparent Zn absorption was similar between treatments. Manganese apparent absorption was greater for the by-product diets compared with the forage diets (16 vs. 11%). A fiber by mineral interaction was observed for Cu apparent absorption, as cows fed hydroxy minerals with forage diets had greater apparent absorption compared with cows fed sulfate minerals; however, the opposite was observed with the by-product diets. Source of supplemental trace minerals and type of fiber in diets affected availability of Cu and Mn and should be considered in ration formulation.
Sigma-Aldrich
Hemoglobin Assay Kit, sufficient for 250 colorimetric tests