Merck
  • Suppression of gut dysbiosis reverses Western diet-induced vascular dysfunction.
Suppression of gut dysbiosis reverses Western diet-induced vascular dysfunction.
American journal of physiology. Endocrinology and metabolism (2018-01-20)
Micah L Battson, Dustin M Lee, Dillon K Jarrell, Shuofei Hou, Kayl E Ecton, Tiffany L Weir, Christopher L Gentile
ABSTRACT
Vascular dysfunction represents a critical preclinical step in the development of cardiovascular disease. We examined the role of the gut microbiota in the development of obesity-related vascular dysfunction. Male C57BL/6J mice were fed either a standard diet (SD) ( n = 12) or Western diet (WD) ( n = 24) for 5 mo, after which time WD mice were randomized to receive either unsupplemented drinking water or water containing a broad-spectrum antibiotic cocktail (WD + Abx) ( n = 12/group) for 2 mo. Seven months of WD caused gut dysbiosis, increased arterial stiffness (SD 412.0 ± 6.0 vs. WD 458.3 ± 9.0 cm/s, P < 0.05) and endothelial dysfunction (28% decrease in max dilation, P < 0.05), and reduced l-NAME-inhibited dilation. Vascular dysfunction was accompanied by significant increases in circulating LPS-binding protein (LBP) (SD 5.26 ± 0.23 vs. WD 11 ± 0.86 µg/ml, P < 0.05) and interleukin-6 (IL-6) (SD 3.27 ± 0.25 vs. WD 7.09 ± 1.07 pg/ml, P < 0.05); aortic expression of phosphorylated nuclear factor-κB (p-NF-κB) ( P < 0.05); and perivascular adipose expression of NADPH oxidase subunit p67phox ( P < 0.05). Impairments in vascular function correlated with reductions in Bifidobacterium spp. Antibiotic treatment successfully abrogated the gut microbiota and reversed WD-induced arterial stiffness and endothelial dysfunction. These improvements were accompanied by significant reductions in LBP, IL-6, p-NF-κB, and advanced glycation end products (AGEs), and were independent from changes in body weight and glucose tolerance. These results indicate that gut dysbiosis contributes to the development of WD-induced vascular dysfunction, and identify the gut microbiota as a novel therapeutic target for obesity-related vascular abnormalities.
Product Description
Millipore
MILLIPLEX® Mouse Adipokine Magnetic Bead Panel - Endocrine Multiplex Assay, The analytes available for this multiplex kit are: IL-6, Insulin, Leptin, MCP-1, PAI-1 (Total), Resistin, TNF-α.