Merck
  • Irreversible inhibition of dihydrodipicolinate synthase by 4-oxo-heptenedioic acid analogues.
Irreversible inhibition of dihydrodipicolinate synthase by 4-oxo-heptenedioic acid analogues.
Bioorganic & medicinal chemistry (2008-11-04)
Berin A Boughton, Michael D W Griffin, Paul A O'Donnell, Renwick C J Dobson, Matthew A Perugini, Juliet A Gerrard, Craig A Hutton
ABSTRACT
We report the synthesis of (2E,5E)-4-oxoheptadienedioic acid and (2E)-4-oxoheptenedioic acid and evaluation of both diester and diacid analogues as inhibitors of bacterial dihydrodipicolinate synthase. Enzyme kinetic studies allowed the determination of second-order rate constants of inactivation; and substrate co-incubation studies have shown the inhibitors act at the active-site. Mass spectrometric analyses have further explored the enzyme-inhibitor interaction and determined the sites of enzyme alkylation.
Sigma-Aldrich
2-Ketobutyric acid, 97%
Sigma-Aldrich
2-Oxovaleric acid, ≥98.0% (T)
Sigma-Aldrich
2-Oxobutyric acid, ≥95%, FG