Merck
  • 6alpha-ethyl-chenodeoxycholic acid (6-ECDCA), a potent and selective FXR agonist endowed with anticholestatic activity.
6alpha-ethyl-chenodeoxycholic acid (6-ECDCA), a potent and selective FXR agonist endowed with anticholestatic activity.
Journal of medicinal chemistry (2002-08-09)
Roberto Pellicciari, Stefano Fiorucci, Emidio Camaioni, Carlo Clerici, Gabriele Costantino, Patrick R Maloney, Antonio Morelli, Derek J Parks, Timothy M Willson
ABSTRACT
A series of 6alpha-alkyl-substituted analogues of chenodeoxycholic acid (CDCA) were synthesized and evaluated as potential farnesoid X receptor (FXR) ligands. Among them, 6alpha-ethyl-chenodeoxycholic acid (6-ECDCA) was shown to be a very potent and selective FXR agonist (EC(50) = 99 nM) and to be endowed with anticholeretic activity in an in vivo rat model of cholestasis.
Sigma-Aldrich
Obeticholic acid, ≥98% (HPLC)