Merck
  • Preparative scale purification of shidasterone, 2-deoxy-polypodine b and 9a,20-dihydroxyecdysone from Silene italica ssp. nemoralis.
Preparative scale purification of shidasterone, 2-deoxy-polypodine b and 9a,20-dihydroxyecdysone from Silene italica ssp. nemoralis.
Journal of chromatographic science (2004-06-11)
Mária Báthori, Zita Pongrácz, Róbert Omacht, Imre Máthé
ABSTRACT
A suitable combination of preparative scale separation methods results in effective clean-up of the ecdysteroids of Silene italica ssp. nemoralis (Waldst. and Kit.) Nyman. The isolation of minor ecdysteroids from the partially purified extract is based on the use of both droplet counter-current chromatography and low-pressure reversed-phase liquid chromatography. The purification is completed by preparative thin-layer chromatography and preparative high-performance liquid chromatography to obtain the minor ecdysteroids, such as 2-deoxy-20-hydroxyecdysone, shidasterone, 2-deoxy-polypodine B, makisterone C, and 9alpha,20-dihydroxyecdysone.
Sigma-Aldrich
Anti-KDR/Flk-1/VEGFR2 Antibody, clone CH-11, clone CH-11, Upstate®, from mouse