Merck
  • Kinetic Study of Glucosamine Production Using Aspergillus sydowii BCRC 31742 under Solid-State Fermentation.
Kinetic Study of Glucosamine Production Using Aspergillus sydowii BCRC 31742 under Solid-State Fermentation.
Molecules (Basel, Switzerland) (2020-10-24)
Jia Wei Peng, Ho Shing Wu
ABSTRACT
In the present study, we aimed to obtain a high yield and productivity for glucosamine using a low-cost solid-state culture with Aspergillus sydowii BCRC 31742. The fermentation conditions, such as inoculum biomass, moisture content, and supplemental volume and mineral salt, were chosen to achieve high productivity of glucosamine (GlcN). When the initial supplemental volume used was 3 mL/g substrate, the yield and productivity of GlcN were 48.7 mg/gds and 0.69 mg/gds·h, respectively. This result will be helpful for the industrialization of the process.
Sigma-Aldrich
Suplatast tosylate, ≥98% (HPLC)