Merck
  • Synthesis of an anti-cariogenic experimental dental composite containing novel drug-decorated copper particles.
Synthesis of an anti-cariogenic experimental dental composite containing novel drug-decorated copper particles.
Materials science & engineering. C, Materials for biological applications (2020-10-01)
Mehwish Pasha, Nawshad Muhammad, Maleeha Nayyer, Jaffar Hussain Bokhari, Hina Ashraf, Sher Zaman Safi, Muhammad Kaleem
ABSTRACT
Secondary caries is one of the most major cause for re-placement of dental composite restorations. Targeting the survival of cariogenic bacteria residing on the restoration surface may reduce this problem. The present study aims to evaluate the antibacterial potential as well as assess the physical and chemical properties of experimental dental composites containing novel drug-decorated copper particles (DDCP) as adjunct antibacterial filler particles. These were incorporated at concentrations of 0%, 0.5%, 0.10%, 0.20%, and 0.25% (w/w) into experimental composite consisting of methacrylate monomers and silanized silica fillers. RESULTS: Direct contact test revealed that the anti-cariogenic potential of experimental composites was more than the control groups. The cell viability assay showed no toxic effect on MC3T3-E1 cell lines in the MTT assay. The microhardness of experimental composites increased as the percentage of DDCP increased, however, the degree of cure was increased only up till the concentration of 0.20%. The release kinetics of the composites reveals that even after 28 days there was a steady and slow release of copper particles signifying the sustained anti-cariogenic effect. CONCLUSION: The experimental composites have good anti-cariogenic potential, which was sustained for one month without any deleterious effect on the physical and chemical properties of resin dental composites.
Sigma-Aldrich
Bisphenol A glycerolate dimethacrylate, glycerol/phenol 1
Sigma-Aldrich
Diurethane dimethacrylate, mixture of isomers, contains 225 ppm±25 ppm BHT as inhibitor, ≥97%