Merck
  • The Cytokine TGF-β Induces Interleukin-31 Expression from Dermal Dendritic Cells to Activate Sensory Neurons and Stimulate Wound Itching.
The Cytokine TGF-β Induces Interleukin-31 Expression from Dermal Dendritic Cells to Activate Sensory Neurons and Stimulate Wound Itching.
Immunity (2020-07-17)
Junji Xu, Peter Zanvit, Lei Hu, Pang-Yen Tseng, Na Liu, Fu Wang, Ousheng Liu, Dunfang Zhang, Wenwen Jin, Nancy Guo, Yichen Han, Jessica Yin, Alexander Cain, Mark A Hoon, Songlin Wang, WanJun Chen
ABSTRACT
Cutaneous wound healing is associated with the unpleasant sensation of itching. Here we investigated the mechanisms underlying this type of itch, focusing on the contribution of soluble factors released during healing. We found high amounts of interleukin 31 (IL-31) in skin wound tissue during the peak of itch responses. Il31-/- mice lacked wound-induced itch responses. IL-31 was released by dermal conventional type 2 dendritic cells (cDC2s) recruited to wounds and increased itch sensory neuron sensitivity. Transfer of cDC2s isolated from late-stage wounds into healthy skin was sufficient to induce itching in a manner dependent on IL-31 expression. Addition of the cytokine TGF-β1, which promotes wound healing, to dermal DCs in vitro was sufficient to induce Il31 expression, and Tgfbr1f/f CD11c-Cre mice exhibited reduced scratching and decreased Il31 expression in wounds in vivo. Thus, cDC2s promote itching during skin would healing via a TGF-β-IL-31 axis with implications for treatment of wound itching.
Sigma-Aldrich
Capsaicin, from Capsicum sp., ≥50% (HPLC)
Sigma-Aldrich
Human IL-31 ELISA Kit, for serum, plasma, cell culture supernatant and urine
Sigma-Aldrich
S3I-201, ≥97% (HPLC)
Sigma-Aldrich
SIS3, ≥98% (HPLC), powder