Merck
  • CACNA1S haploinsufficiency confers resistance to New World arenavirus infection.
CACNA1S haploinsufficiency confers resistance to New World arenavirus infection.
Proceedings of the National Academy of Sciences of the United States of America (2020-07-29)
Nicolás Sarute, Susan R Ross
ABSTRACT
Understanding the genetics of susceptibility to infectious agents is of great importance to our ability to combat disease. Here, we show that voltage-gated calcium channels (VGCCs) are critical for cellular binding and entry of the New World arenaviruses Junín and Tacaribe virus, suggesting that zoonosis via these receptors could occur. Moreover, we demonstrate that α1s haploinsufficiency renders cells and mice more resistant to infection by these viruses. In addition to being more resistant to infection, haploinsufficient cells and mice required a lower dosage of VGCC antagonists to block infection. These studies underscore the importance of genetic variation in susceptibility to both viruses and pharmaceutics.
Sigma-Aldrich
MISSION® esiRNA, targeting human UGT1A3
Pricing and availability is not currently available.