Merck
  • Blocking ACAT-1 Activity for Tumor Therapy with Fluorescent Hyperstar Polymer-Encapsulated Avasimible.
Blocking ACAT-1 Activity for Tumor Therapy with Fluorescent Hyperstar Polymer-Encapsulated Avasimible.
Macromolecular bioscience (2020-05-15)
Ting Bai, Bobo Zhu, Dongyan Shao, Ziyang Lian, Pei Liu, Junling Shi, Jie Kong
ABSTRACT
Targeting the distinct cholesterol metabolism of tumor cells is proposed as a novel way to treat tumors. Blocking acyl-CoA cholesterol acyltransferase-1 (ACAT-1) by the inhibitor avasimible (Ava), which elevates intracellular free cholesterol levels, is shown to effectively induce apoptosis. However, Ava faces disadvantages of poor water solubility, a short half-life, and no capability for fluorescence detection, which have greatly limited its application. Herein, a fluorescent hyperstar polymer (FHSP) is developed to encapsulate Ava to improve its ability to inhibit HeLa cells and K562 cells. The results of this study show that the obtained Ava-FHSP micelles possess a high drug loading capacity of 22.7% and bright green fluorescence. Ava and Ava-FHSP are cytotoxic to both HeLa and K562 cells and cause reductions in cell size, nuclear lysis, and chromatin condensation and hindered proliferation of both cell types by causing S phase cell cycle arrest. Further mechanistic analysis indicates that Ava-FHSP reduces the protein and messenger RNA expression of ACAT-1 and significantly increases intracellular free cholesterol levels, which can increase endoplasmic reticulum stress and finally cause cell apoptosis. All these results suggest that this fluorescent hyperstar polymer represents a potential therapeutic tumor strategy by changing the cholesterol metabolism of tumor cells.
Sigma-Aldrich
2,3-Dibromomaleimide, 97%