Merck
  • Differentiation of Cosmetic Foundation Creams Using Attenuated Total Reflection Fourier-Transform Infrared Spectroscopy: A Rapid and NonDestructive Approach in Trace Evidence Analysis.
Differentiation of Cosmetic Foundation Creams Using Attenuated Total Reflection Fourier-Transform Infrared Spectroscopy: A Rapid and NonDestructive Approach in Trace Evidence Analysis.
Journal of forensic sciences (2019-12-14)
Sweety Sharma, Rito Chophi, Harkamalpreet Kaur, Rajinder Singh
ABSTRACT
Cosmetic foundation creams are encountered as evidentiary material in criminal investigations, particularly in cases related to sexual and physical assault against women. Analyzing foundation cream exhibit is a challenging task as the exhibit is recovered in trace quantity with similar hue. In the present study, ATR FTIR spectroscopy which is a rapid, nondestructive, sensitive, reliable, and safe alternative to other analytical techniques has been used to differentiate 31 cosmetic foundation creams belonging to 23 different brands supported by chemometric methods such as principal component analysis (PCA) and linear discriminant analysis (LDA). The discriminating power of visual analysis is found to be 98.0%, while PCA and LDA further increased the discriminating power to 99.3% and 100%, respectively. The blind test is conducted with three unknown samples (pretended to be unknown to the analyst), which were correctly linked with their respective source. Further, the effect of substrate such as tissue paper (dry and wet) and white cotton cloth during sample analysis are also examined to simulate the actual forensic casework conditions. The stains on substrates could be identified and linked to its parent product as well. The reported method provides significant results for the differentiation of cosmetic foundation creams.
Sigma-Aldrich
L-Alanine-2-d, ≥98 atom % D, ≥98% (CP)