Merck
  • Conductive Metal-Organic Frameworks with Extra Metallic Sites as an Efficient Electrocatalyst for the Hydrogen Evolution Reaction.
Conductive Metal-Organic Frameworks with Extra Metallic Sites as an Efficient Electrocatalyst for the Hydrogen Evolution Reaction.
Advanced science (Weinheim, Baden-Wurttemberg, Germany) (2020-05-10)
Hao Huang, Yue Zhao, Yimin Bai, Fumin Li, Ying Zhang, Yu Chen
ABSTRACT
The 2D conductive metal-organic frameworks (MOFs) are expected to be an ideal electrocatalyst due to their high utilization of metal atoms. Exploring a new conjugated ligand with extra active metallic center can further boost the structural advantages of conductive MOFs. In this work, hexaiminohexaazatrinaphthalene (HAHATN) is employed as a conjugated ligand to construct bimetallic sited conductive MOFs (M23(M13∙HAHATN)2) with an extra M-N2 moiety. Density functional theory (DFT) calculations demonstrate that the 2D conjugated framework renders M23(M13∙HAHATN)2 a high electric conductivity with narrow bandgap (0.19 eV) for electron transfer and a favorable in-plane porous structure (2.7 nm) for mass transfer. Moreover, the metal atom at the extra M-N2 moiety has a higher unsaturation degree than that at M-N4 linkage, resulting in a stronger ability to donate electrons for enhancing electroactivity. These characteristics endow the new conductive MOFs with an enhanced electroactivity for hydrogen evolution reaction (HER) electrocatalysis. Among the series of M23(M13∙HAHATN)2 MOF, Ni3(Ni3∙HAHATN)2 nanosheets with the optimal structure exhibit a small overpotential of 115 mV at 10 mA cm-2, low Tafel slope of (45.6 mV dec-1), and promising electrocatalytic stability for HER. This work provides an effective strategy for designing conductive MOFs with a favorable structure for electrocatalysis.
Sigma-Aldrich
1,2,4,5-Benzenetetramine tetrahydrochloride, ≥95%
Sigma-Aldrich
Hexaketocyclohexane octahydrate, 97%