Merck
  • Formation of odorous aldehydes, nitriles and N-chloroaldimines from combined leucine in short oligopeptides during chlorination.
Formation of odorous aldehydes, nitriles and N-chloroaldimines from combined leucine in short oligopeptides during chlorination.
Water research (2020-04-18)
Luyang Cai, Lei Li, Shuili Yu
ABSTRACT
Previous studies have focused on investigating the formation of odorous by-products during the chlorination of free amino acids (AAs). However, studies on the formation of odorous by-products during the chlorination of combined AAs, which are much more abundant in natural waters than free AAs, are very limited. In this study, the generation of odorous aldehyde, nitrile and N-chloroaldimine from short oligopeptides containing combined Leucine (Leu) (a typical precursor of odorous by-products), including glycylleucine (Gly-Leu), leucylglycine (Leu-Gly), and trileucine (Leu-Leu-Leu), was investigated. The reaction mechanisms were then proposed based on Acquity UPLC-qTOF mass spectrometer measurement and kinetic studies modelled with Kintecus. The results indicated that a series of sequential reactions, including substitution, dehydrohalogenation, β-elimination, hydrolysis and decarboxylation reactions, occurred during the chlorination of short oligopeptides. The chlorination of Gly-Leu and Leu-Leu-Leu formed free Leu, which continued to react with chlorine, producing isovaleraldehyde, isovaleronitrile and N-chloroisovaleraldimine. Compared with Gly-Leu, Leu-Leu-Leu produced less free Leu, and therefore, a smaller amount of Leu-derived odorous by-products was generated. Leu-Gly produced free Gly, which was not a precursor of odorous by-products. Thus, neither isovaleraldehyde nor N-chloroisovaleraldimine was formed. Notably, isovaleronitriles can be formed directly from a β-elimination reaction during chlorination of Leu-Gly and Leu-Leu-Leu, and thus high yields of isovaleronitriles were observed after chlorination. The yields of odorous by-products during chlorination of short oligopeptides increased with increasing Cl/N ratios (the molar ratio of chlorine to nitrogen in the AAs) and reached their maximum at Cl/N = 2.4, except the yield of isovaleraldehyde formed from Gly-Leu reached its maximum at Cl/N = 1.6. UV and UV/H2O2 pre-treatments decreased odorous by-product formation during subsequent chlorination through non-peptide bond breaking of short oligopeptides. This study facilitates the identification of the causes of off-flavour problems in drinking water and the development of ways to control these problems.
Sigma-Aldrich
Isovaleronitrile, 98%