Merck
  • Synthesis and Properties of pH-Thermo Dual Responsive Semi-IPN Hydrogels Based on N,N'-Diethylacrylamide and Itaconamic Acid.
Synthesis and Properties of pH-Thermo Dual Responsive Semi-IPN Hydrogels Based on N,N'-Diethylacrylamide and Itaconamic Acid.
Polymers (2020-05-21)
Huynh Nguyen Anh Tuan, Vo Thi Thu Nhu
ABSTRACT
A series of semi-interpenetrating polymer network (semi-IPN) hydrogels based on N,N'-diethylacrylamide (DEA) and itaconamic acid (IAM) were synthesized by changing the molar ratio of linear copolymer P(DEA-co-IAM) and DEA monomer. Linear copolymer P(DEA-co-IAM) was introduced into a solution of DEA monomer to prepare pH-thermo dual responsive P(DEA-co-IAM)/PDEA semi-IPN hydrogels. The thermal gravimetric analysis (TGA) revealed that the semi-IPN hydrogel has a higher thermal stability than the conventional hydrogel, while the interior morphology by scanning electron microscopy (SEM) showed a porous structure with the pore sizes could be controlled by changing the ratio of linear copolymer in the obtained hydrogels. The oscillatory parallel-plate rheological measurements and compression tests demonstrated a viscoelastic behavior and superior mechanical properties of the semi-IPN hydrogels. Besides, the lower critical solution temperature (LCST) of the linear copolymers increased with the increase of IAM content in the feed, while the semi-IPN hydrogels increased LCSTs with the increase of linear copolymer content introduced. The pH-thermo dual responsive of the hydrogels was investigated using the swelling behavior in various pH and temperature conditions. Finally, the swelling and deswelling rate of the hydrogels were also studied. The results indicated that the pH-thermo dual responsive semi-IPN hydrogels were synthesized successfully and may be a potential material for biomedical, drug delivery or absorption applications. The further applications of semi-IPN hydrogels are being conducted.
Sigma-Aldrich
N,N-Diethylacrylamide, contains <200 ppm MEHQ as inhibitor, 99%