Merck
  • The prophylactic and therapeutic activity of a broadly active ribonucleoside analog in a murine model of intranasal venezuelan equine encephalitis virus infection.
The prophylactic and therapeutic activity of a broadly active ribonucleoside analog in a murine model of intranasal venezuelan equine encephalitis virus infection.
Antiviral research (2019-09-09)
George R Painter, Richard A Bowen, Gregory R Bluemling, John DeBergh, Vindhya Edpuganti, Prabhakar R Gruddanti, David B Guthrie, Michael Hager, Damien L Kuiper, Mark A Lockwood, Deborah G Mitchell, Michael G Natchus, Zachary M Sticher, Alexander A Kolykhalov
ABSTRACT
The New World alphaviruses Venezuelan, Eastern, and Western equine encephalitis viruses (VEEV, EEEV and WEEV, respectively) commonly cause a febrile disease that can progress to meningoencephalitis, resulting in significant morbidity and mortality. To address the need for a therapeutic agent for the treatment of Alphavirus infections, we identified and pursued preclinical characterization of a ribonucleoside analog EIDD-1931 (β-D-N4-hydroxycytidine, NHC), which has shown broad activity against alphaviruses in vitro and has a very high genetic barrier for development of resistance. To be truly effective as a therapeutic agent for VEEV infection a drug must penetrate the blood brain barrier and arrest virus replication in the brain. High plasma levels of EIDD-1931 are rapidly achieved in mice after oral dosing. Once in the plasma EIDD-1931 is efficiently distributed into organs, including brain, where it is rapidly converted to its active 5'-triphosphate. EIDD-1931 showed a good safety profile in mice after 7-day repeated dosing with up to 1000 mg/kg/day doses. In mouse model studies, EIDD-1931 was 90-100% effective in protecting mice against lethal intranasal infection when therapeutic treatment was started as late as 24 h post-infection, and partial protection was achieved when treatment was delayed for 48 h post-infection. These results support further preclinical development of EIDD-1931 as a potential anti-alphavirus drug.
Sigma-Aldrich
EIDD-1931, ≥98% (HPLC)