- Staudinger ligation of alpha-azido acids retains stereochemistry.
Staudinger ligation of alpha-azido acids retains stereochemistry.
The Journal of organic chemistry (2002-07-06)
Matthew B Soellner, Bradley L Nilsson, Ronald T Raines
PMID12098322
ABSTRACT
The Staudinger ligation of peptides with a C-terminal phosphinothioester and N-terminal azide is an emerging method in protein chemistry. Here, the first Staudinger ligations of nonglycyl azides are reported and shown to proceed both in nearly quantitative yield and with no detectable effect on the stereochemistry at the alpha-carbon of the azide. These results demonstrate further the potential of the Staudinger ligation as a general method for the total synthesis of proteins from peptide fragments.