Merck
  • miR‑508‑3p suppresses the development of ovarian carcinoma by targeting CCNA2 and MMP7.
miR‑508‑3p suppresses the development of ovarian carcinoma by targeting CCNA2 and MMP7.
International journal of oncology (2020-05-08)
Fei Guo, Kai Zhang, Meiyue Li, Lei Cui, Guoyan Liu, Ye Yan, Wenyan Tian, Fei Teng, Yanfang Zhang, Chao Gao, Jinping Gao, Yingmei Wang, Fengxia Xue
ABSTRACT
Ovarian cancer is the most lethal gynecological tumor, and the 5‑year survival rate is only ~40%. The poor survival rate is due to cancer diagnosis at an advanced stage, when the tumor has metastasized. A better understanding of the molecular pathogenesis of tumor growth and metastasis is needed to improve patient prognosis. MicroRNAs (miRs) regulate carcinogenesis and development of cancers. However, the role of miR‑508‑3p in ovarian cancer remains largely unknown. Thus, the present study aimed to investigate the possible functions of miR‑508‑3p in the modulation of development of ovarian cancer. The results of the present study demonstrated that miR‑508‑3p mimics inhibited ovarian cancer cell proliferation, migration and invasion. Reporter gene assay results demonstrated that miR‑508‑3p suppressed cancer cell proliferation by directly targeting the 3'‑untranslated region (UTR) of cyclin A2 (CCNA2) and suppressed migration and invasion by directly targeting the 3'‑UTR of matrix metalloproteinase 7 (MMP7). In addition, high CCNA2 and MMP7 expression levels were associated with low miR‑508‑3p expression in ovarian cancer tissues. Furthermore, miR‑508‑3p and CCNA2 were independent predictors for overall survival in patients with ovarian cancer. To the best of our knowledge, this is the first study to demonstrated that miR‑508‑3p suppressed ovarian cancer development by directly targeting CCNA2 and MMP7. The results of this study suggested the potential value of miR‑508‑3p and CCNA2 as prognostic indicators and therapeutics for ovarian cancer.
Sigma-Aldrich
MISSION® esiRNA, targeting human CCNA2
Roche
Anti-Digoxigenin-AP, Fab fragments, from sheep