Merck
  • The scaffold protein p62 regulates adaptive thermogenesis through ATF2 nuclear target activation.
The scaffold protein p62 regulates adaptive thermogenesis through ATF2 nuclear target activation.
Nature communications (2020-05-10)
Katrin Fischer, Anna Fenzl, Dianxin Liu, Kenneth A Dyar, Maximilian Kleinert, Markus Brielmeier, Christoffer Clemmensen, Anna Fedl, Brian Finan, Andre Gessner, Martin Jastroch, Jianfeng Huang, Susanne Keipert, Martin Klingenspor, Jens C Brüning, Manfred Kneilling, Florian C Maier, Ahmed E Othman, Bernd J Pichler, Ines Pramme-Steinwachs, Stephan Sachs, Angelika Scheideler, Wolfgang M Thaiss, Henriette Uhlenhaut, Siegfried Ussar, Stephen C Woods, Julia Zorn, Kerstin Stemmer, Sheila Collins, Maria Diaz-Meco, Jorge Moscat, Matthias H Tschöp, Timo D Müller
ABSTRACT
During β-adrenergic stimulation of brown adipose tissue (BAT), p38 phosphorylates the activating transcription factor 2 (ATF2) which then translocates to the nucleus to activate the expression of Ucp1 and Pgc-1α. The mechanisms underlying ATF2 target activation are unknown. Here we demonstrate that p62 (Sqstm1) binds to ATF2 to orchestrate activation of the Ucp1 enhancer and Pgc-1α promoter. P62Δ69-251 mice show reduced expression of Ucp1 and Pgc-1α with impaired ATF2 genomic binding. Modulation of Ucp1 and Pgc-1α expression through p62 regulation of ATF2 signaling is demonstrated in vitro and in vivo in p62Δ69-251 mice, global p62-/- and Ucp1-Cre p62flx/flx mice. BAT dysfunction resulting from p62 deficiency is manifest after birth and obesity subsequently develops despite normal food intake, intestinal nutrient absorption and locomotor activity. In summary, our data identify p62 as a master regulator of BAT function in that it controls the Ucp1 pathway through regulation of ATF2 genomic binding.
Sigma-Aldrich
Monoclonal ANTI-FLAG® M2 antibody produced in mouse, 1 mg/mL, clone M2, affinity isolated antibody, buffered aqueous solution (50% glycerol, 10 mM sodium phosphate, and 150 mM NaCl, pH 7.4)
Sigma-Aldrich
Anti-Rabbit IgG (whole molecule)–Alkaline Phosphatase antibody produced in goat, affinity isolated antibody, buffered aqueous glycerol solution
Sigma-Aldrich
Anti-Mouse IgG (whole molecule)−Alkaline Phosphatase antibody produced in goat, affinity isolated antibody, buffered aqueous glycerol solution